Learn More
ABS Undercounter Built-In Pharmacy Refrigerator
Experience the best in vaccine refrigeration, certified to the NSF/ANSI 456 Vaccine Standard.
Supplier:  ABS PHABTNSFUCBI0404
 View more versions of this product
Catalog No. 361058615
Only null left
Description
Description
These cutting-edge 130 L pharmacy refrigerators are certified in accordance with the NSF/ANSI 456 Standard for Vaccine Storage. With this certification, units protect pharmaceuticals at optimal temperatures, preventing waste and allowing for peak delivery.
  • These built-in refrigerators utilize microprocessor controllers and feature temperature alarms, remote alarm contacts, and probe access ports with included probes.
  • Built-in units are designed with front ventilation systems, allowing easy installation into a bench or countertop.
  • All units run on natural, hydrocarbon refrigerant for environmental health and energy efficiency.
Specifications
Chemical Identifiers
Vaccine Refrigerator
130 L
Solid Swing
3
Powder-coated Steel
Microprocessor Temperature Controller
NEMA 5-15
4.6 cu. ft.
1°C to 10°C
1
Scratch-resistant Painted Steel
Undercounter
H-drive
115 V
Specifications
Vaccine Refrigerator
130 L
Solid Swing
3
Powder-coated Steel
Undercounter
H-drive
115 V
4.6 cu. ft.
1°C to 10°C
1
Scratch-resistant Painted Steel
cULus, ETL
Microprocessor Temperature Controller
NEMA 5-15
Product Suggestions
Product Suggestions