missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Undecylenic acid, 98+%

CAS: 112-38-9 | C11H20O2 | 184.28 g/mol

Supplier:  Thermo Scientific Chemicals 154495000

 View more versions of this product

Catalog No. AC154495000


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Undecylenic acid
112-38-9
Yellow
275.0°C
Authentic
Glass bottle
1.4470 to 1.4480
MFCD00004442
0.911
10-undecenoic acid, undecylenic acid, desenex, 10-hendecenoic acid, renselin, declid, sevinon, 10-undecylenic acid, 9-undecylenic acid, undecyl-10-enic acid
FRPZMMHWLSIFAZ-UHFFFAOYSA-N
undec-10-enoic acid
5634
184.28
Crystalline Low Melting Solid or Liquid
99%
24.5°C
0.9110g/mL
148°C
97% min. (GC)
C11H20O2
H2C=CH(CH2)8CO2H
500 g
15, 10033
Solubility in water: insoluble. Other solubilities: miscible with alcohol,chloroform,ether,benzene,with fixed and volatile oils
C=CCCCCCCCCC(=O)O
184.28
CHEBI:35045
99%

Chemical Identifiers

112-38-9
184.28
FRPZMMHWLSIFAZ-UHFFFAOYSA-N
5634
undec-10-enoic acid
C11H20O2
MFCD00004442
10-undecenoic acid, undecylenic acid, desenex, 10-hendecenoic acid, renselin, declid, sevinon, 10-undecylenic acid, 9-undecylenic acid, undecyl-10-enic acid
CHEBI:35045
C=CCCCCCCCCC(=O)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful to aquatic life with long lasting effects.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 203-965-8

RTECSNumber : YQ2975000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only