missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Undecylenic acid, 98+%

Catalog No. p-210623
Change view
Click to view available options
Quantity:
250 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC154492500 250 g Glass bottle
AC154495000 500 g Glass bottle
AC154490025 2.5 kg Plastic drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC154492500 Supplier Thermo Scientific Chemicals Supplier No. 154492500
Only null left

CAS: 112-38-9 | C11H20O2 | 184.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 112-38-9
Molecular Formula C11H20O2
Molecular Weight (g/mol) 184.28
MDL Number MFCD00004442
InChI Key FRPZMMHWLSIFAZ-UHFFFAOYSA-N
Synonym 10-undecenoic acid, undecylenic acid, desenex, 10-hendecenoic acid, renselin, declid, sevinon, 10-undecylenic acid, 9-undecylenic acid, undecyl-10-enic acid
PubChem CID 5634
ChEBI CHEBI:35045
IUPAC Name undec-10-enoic acid
SMILES C=CCCCCCCCCC(=O)O

Specifications

Melting Point 24.5°C
Color Yellow
Density 0.9110g/mL
Boiling Point 275.0°C
Flash Point 148°C
Infrared Spectrum Authentic
Assay Percent Range 97% min. (GC)
Packaging Glass bottle
Linear Formula H2C=CH(CH2)8CO2H
Refractive Index 1.4470 to 1.4480
Quantity 250 g
Specific Gravity 0.911
Merck Index 15, 10033
Solubility Information Solubility in water: insoluble. Other solubilities: miscible with alcohol,chloroform,ether,benzene,with fixed and volatile oils
Formula Weight 184.28
Percent Purity 99%
Physical Form Crystalline Low Melting Solid or Liquid
Chemical Name or Material Undecylenic acid, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful to aquatic life with long lasting effects.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 203-965-8
RTECSNumber YQ2975000
TSCA TSCA

RUO – Research Use Only