missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Undecanoyl Chloride 97.0+%, TCI America™
$92.41 - $177.11
Chemical Identifiers
| CAS | 17746-05-3 |
|---|---|
| Molecular Formula | C11H21ClO |
| Molecular Weight (g/mol) | 204.738 |
| MDL Number | MFCD00000739 |
| InChI Key | JUKPJGZUFHCZQI-UHFFFAOYSA-N |
| Synonym | n-undecanoyl chloride, undecanoic acid chloride, undecanoylchloride, n-undecanoylchloride, undecanoyl chloride, acmc-1c80a, ksc175q6f |
| PubChem CID | 87287 |
| IUPAC Name | undecanoyl chloride |
| SMILES | CCCCCCCCCCC(=O)Cl |
Chemical Identifiers
| 17746-05-3 | |
| 204.738 | |
| JUKPJGZUFHCZQI-UHFFFAOYSA-N | |
| 87287 | |
| CCCCCCCCCCC(=O)Cl |
| C11H21ClO | |
| MFCD00000739 | |
| n-undecanoyl chloride, undecanoic acid chloride, undecanoylchloride, n-undecanoylchloride, undecanoyl chloride, acmc-1c80a, ksc175q6f | |
| undecanoyl chloride |
Specifications
| 17746-05-3 | |
| 130°C | |
| MFCD00000739 | |
| 3265 | |
| JUKPJGZUFHCZQI-UHFFFAOYSA-N | |
| undecanoyl chloride | |
| 87287 | |
| ≥97.0% (GC,T) | |
| Undecanoyl Chloride |
| Yellow | |
| C11H21ClO | |
| 10 mL | |
| n-undecanoyl chloride, undecanoic acid chloride, undecanoylchloride, n-undecanoylchloride, undecanoyl chloride, acmc-1c80a, ksc175q6f | |
| CCCCCCCCCCC(=O)Cl | |
| 204.738 | |
| 204.74 | |
| Liquid |
Safety and Handling
TSCA : Yes