missing translation for 'onlineSavingsMsg'
Learn More

Undecanoic acid, ≥99% (GC), MilliporeSigma™ Supelco™

Catalog No. 111012488
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012488 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012488 Supplier Merck Emd Millipore Supplier No. 897641GF

Undecanoic acid is a medium chain fatty acid and the most fungitoxic compound commonly used in the treatment of superficial mycoses in humans.

Specifications

CAS 112-37-8
Flash Point 113°C
Shipping Condition Ambient
Physical Form Neat
Linear Formula CH3(CH2)9 COOH
MDL Number MFCD00002730
Melting Point 28°C to 31°C
Boiling Point 228°C (160 mmHg); 248°C to 250°C
Synonym Hendecanoic acid
Type Analytical Standard
Quantity 1 g
Formula Weight 186.29
Percent Purity ≥99% (GC)
Grade Analytical Standard
Show More Show Less