missing translation for 'onlineSavingsMsg'
Learn More

Undecanenitrile 97.0+%, TCI America™

Catalog No. U004425G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
U004425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. U004425G Supplier TCI America Supplier No. U004425G
Only null left

Chemical Identifiers

CAS 2244-07-7
Molecular Formula C11H21N
Molecular Weight (g/mol) 167.30
MDL Number MFCD00001945
InChI Key SZKKNEOUHLFYNA-UHFFFAOYSA-N
Synonym Decyl Cyanide
PubChem CID 16722
IUPAC Name undecanenitrile
SMILES CCCCCCCCCCC#N

Specifications

Color Yellow
Boiling Point 129°C
Quantity 25 g
Formula Weight 167.30
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Undecanenitrile
EINECSNumber (2)-1507
TSCA Yes