accessibility menu, dialog, popup

UserName

CAS RN 2244-07-7

H3CN

CAS RN 2244-07-7

IUPAC Name: undecanenitrile
Synonyms: undecanenitrile
Molecular Weight (g/mol): 167.3
Molecular Formula: C11H21N
InChi Key: SZKKNEOUHLFYNA-UHFFFAOYSA-N
SMILES: CCCCCCCCCCC#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results