missing translation for 'onlineSavingsMsg'
Learn More

Undecanedinitrile 95.0+%, TCI America™

Catalog No. U00595ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
U00595ML 5 mL
U005925ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. U00595ML Supplier TCI America Supplier No. U00595ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 71172-36-6
Molecular Formula C11H18N2
Molecular Weight (g/mol) 178.279
MDL Number MFCD00039506
InChI Key ISIQQQYKUPBYSL-UHFFFAOYSA-N
Synonym 1,9-Dicyanononane
PubChem CID 247906
IUPAC Name undecanedinitrile
SMILES C(CCCCC#N)CCCCC#N

Specifications

Color Colorless
Boiling Point 190°C
Quantity 5 mL
UN Number 3276
Formula Weight 178.28
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Undecanedinitrile