accessibility menu, dialog, popup

UserName

NN

CAS RN 71172-36-6

IUPAC Name: undecanedinitrile
Synonyms: undecanedinitrile
Molecular Weight (g/mol): 178.28
Molecular Formula: C11H18N2
InChi Key: ISIQQQYKUPBYSL-UHFFFAOYSA-N
SMILES: N#CCCCCCCCCCC#N
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results