missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| Undecane | |
| -26°C | |
| 195°C | |
| MFCD00008959 | |
| 2330 | |
| RSJKGSCJYJTIGS-UHFFFAOYSA-N | |
| undecane | |
| 14257 | |
| 156.31 | |
| Liquid |
| 1120-21-4 | |
| Colorless | |
| C11H24 | |
| 500 mL | |
| n-undecane, hendecane, n-hendecane, decane, methyl, unii-jv0qt00nue, ccris 3796, jv0qt00nue, undecane, undecane, analytical standard, und | |
| CCCCCCCCCCC | |
| 156.31 | |
| CHEBI:46342 | |
| ≥99.0% (GC) |
Chemical Identifiers
| 1120-21-4 | |
| 156.31 | |
| RSJKGSCJYJTIGS-UHFFFAOYSA-N | |
| 14257 | |
| undecane |
| C11H24 | |
| MFCD00008959 | |
| n-undecane, hendecane, n-hendecane, decane, methyl, unii-jv0qt00nue, ccris 3796, jv0qt00nue, undecane, undecane, analytical standard, und | |
| CHEBI:46342 | |
| CCCCCCCCCCC |
Safety and Handling
EINECSNumber : (2)-0010
RTECSNumber : YQ1525000
TSCA : Yes