missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Undecane 99.0+%, TCI America™
$42.87 - $145.40
Chemical Identifiers
| CAS | 1120-21-4 |
|---|---|
| Molecular Formula | C11H24 |
| Molecular Weight (g/mol) | 156.31 |
| MDL Number | MFCD00008959 |
| InChI Key | RSJKGSCJYJTIGS-UHFFFAOYSA-N |
| Synonym | n-undecane, hendecane, n-hendecane, decane, methyl, unii-jv0qt00nue, ccris 3796, jv0qt00nue, undecane, undecane, analytical standard, und |
| PubChem CID | 14257 |
| ChEBI | CHEBI:46342 |
| IUPAC Name | undecane |
| SMILES | CCCCCCCCCCC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
U000225ML
|
TCI America
U000225ML |
25 mL |
Each for $42.87
|
|
|||||
|
U0002500ML
|
TCI America
U0002500ML |
500 mL |
Each for $145.40
|
|
|||||
Chemical Identifiers
| 1120-21-4 | |
| 156.31 | |
| RSJKGSCJYJTIGS-UHFFFAOYSA-N | |
| 14257 | |
| undecane |
| C11H24 | |
| MFCD00008959 | |
| n-undecane, hendecane, n-hendecane, decane, methyl, unii-jv0qt00nue, ccris 3796, jv0qt00nue, undecane, undecane, analytical standard, und | |
| CHEBI:46342 | |
| CCCCCCCCCCC |
Specifications
| 1120-21-4 | |
| Colorless | |
| C11H24 | |
| 25 mL | |
| n-undecane, hendecane, n-hendecane, decane, methyl, unii-jv0qt00nue, ccris 3796, jv0qt00nue, undecane, undecane, analytical standard, und | |
| CCCCCCCCCCC | |
| 156.31 | |
| CHEBI:46342 | |
| ≥99.0% (GC) | |
| Undecane |
| -26°C | |
| 195°C | |
| MFCD00008959 | |
| 2330 | |
| RSJKGSCJYJTIGS-UHFFFAOYSA-N | |
| undecane | |
| 14257 | |
| 156.31 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-0010
RTECSNumber : YQ1525000
TSCA : Yes