missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Undecanal 97.0+%, TCI America™
$78.29 - $307.57
Chemical Identifiers
| CAS | 112-44-7 |
|---|---|
| Molecular Formula | C11H22O |
| Molecular Weight (g/mol) | 170.296 |
| MDL Number | MFCD00007016 |
| InChI Key | KMPQYAYAQWNLME-UHFFFAOYSA-N |
| Synonym | undecanaldehyde, n-undecanal, undecyl aldehyde, undecylic aldehyde, 1-undecanal, undecylaldehyde, hendecanal, hendecanaldehyde, aldehyde c-11, n-undecyl aldehyde |
| PubChem CID | 8186 |
| ChEBI | CHEBI:46202 |
| IUPAC Name | undecanal |
| SMILES | CCCCCCCCCCC=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
U000925ML
|
TCI America
U000925ML |
25 mL |
Each for $78.29
|
|
|||||
|
U0009250ML
|
TCI America
U0009250ML |
250 mL |
Each for $307.57
|
|
|||||
Chemical Identifiers
| 112-44-7 | |
| 170.296 | |
| KMPQYAYAQWNLME-UHFFFAOYSA-N | |
| 8186 | |
| undecanal |
| C11H22O | |
| MFCD00007016 | |
| undecanaldehyde, n-undecanal, undecyl aldehyde, undecylic aldehyde, 1-undecanal, undecylaldehyde, hendecanal, hendecanaldehyde, aldehyde c-11, n-undecyl aldehyde | |
| CHEBI:46202 | |
| CCCCCCCCCCC=O |
Specifications
| 112-44-7 | |
| Yellow | |
| C11H22O | |
| 25 mL | |
| KMPQYAYAQWNLME-UHFFFAOYSA-N | |
| undecanal | |
| 8186 | |
| 170.30 | |
| Liquid |
| -2°C | |
| 80°C | |
| MFCD00007016 | |
| undecanaldehyde, n-undecanal, undecyl aldehyde, undecylic aldehyde, 1-undecanal, undecylaldehyde, hendecanal, hendecanaldehyde, aldehyde c-11, n-undecyl aldehyde | |
| CCCCCCCCCCC=O | |
| 170.296 | |
| CHEBI:46202 | |
| ≥97.0% (GC) | |
| Undecanal |
Safety and Handling
EINECSNumber : (2)-0494
RTECSNumber : YQ1500000
TSCA : Yes