missing translation for 'onlineSavingsMsg'
Learn More

Undecanal 97.0+%, TCI America™

Catalog No. U000925ML
Change view
Click to view available options
Quantity:
25 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
U000925ML 25 mL
U0009250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. U000925ML Supplier TCI America Supplier No. U000925ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 112-44-7
Molecular Formula C11H22O
Molecular Weight (g/mol) 170.296
MDL Number MFCD00007016
InChI Key KMPQYAYAQWNLME-UHFFFAOYSA-N
Synonym undecanaldehyde, n-undecanal, undecyl aldehyde, undecylic aldehyde, 1-undecanal, undecylaldehyde, hendecanal, hendecanaldehyde, aldehyde c-11, n-undecyl aldehyde
PubChem CID 8186
ChEBI CHEBI:46202
IUPAC Name undecanal
SMILES CCCCCCCCCCC=O

Specifications

Melting Point -2°C
Color Yellow
Boiling Point 80°C
Quantity 25 mL
Formula Weight 170.30
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Undecanal
EINECSNumber (2)-0494
RTECSNumber YQ1500000
TSCA Yes