missing translation for 'onlineSavingsMsg'
Learn More

gamma-Undecalactone, 98%

Catalog No. AC259490250
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC259490250 25 mL Glass bottle
AC259491000 100 mL Glass bottle
AC259495000 500 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC259490250 Supplier Thermo Scientific Chemicals Supplier No. 259490250

CAS: 104-67-6 | C11H20O2 | 184.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 104-67-6
Molecular Formula C11H20O2
Molecular Weight (g/mol) 184.28
MDL Number MFCD00005405
InChI Key PHXATPHONSXBIL-UHFFFAOYSA-N
Synonym gamma-undecalactone, 4-undecanolide, 5-heptyldihydrofuran-2 3h-one, persicol, 1,4-undecanolide, peach aldehyde, peach lactone, 2 3h-furanone, 5-heptyldihydro, aldehyde c-14 peach, gamma-undecanolactone
PubChem CID 7714
IUPAC Name 5-heptyloxolan-2-one
SMILES CCCCCCCC1CCC(=O)O1

Specifications

Color Colorless
Density 0.9425g/mL
Boiling Point 286.0°C
Flash Point 145°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4490 to 1.4540
Quantity 25 mL
Specific Gravity 0.9425
Formula Weight 184.28
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material γ-Undecalactone
Show More Show Less
Hazard Statement GHS H Statement
Harmful to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Avoid release to the environment.
Dispose of contents/container to an approved waste disposal plant.
EINECSNumber 203-225-4

RUO – Research Use Only