accessibility menu, dialog, popup

UserName

CAS RN 104-67-6

IUPAC Name: (5S)-5-heptyloxolan-2-one
Synonyms: gamma-undecalactone 1,4-undecanolide gamma-undekalakton gamma-undecalactone (natural) aldehyde C-14 peach peach lactone gamma-undecanolide 1,4-hendecanolide peach aldehyde 4-undecanolide
Molecular Weight (g/mol): 184.28
Molecular Formula: C11H20O2
InChi Key: PHXATPHONSXBIL-JTQLQIEISA-N
SMILES: CCCCCCC[C@H]1CCC(=O)O1
Boiling Point 
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (5)
  • (1)
  • (2)
  • (2)