missing translation for 'onlineSavingsMsg'
Learn More

Undecafluoroiodocyclohexane (stabilized with Copper chip) 98.0+%, TCI America™

Catalog No. U00801G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
U00801G 1 g
U00805G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. U00801G Supplier TCI America Supplier No. U00801G

Chemical Identifiers

CAS 355-69-1
Molecular Formula C6F11I
Molecular Weight (g/mol) 407.953
MDL Number MFCD00156095
InChI Key WYRAAMGCHCLHRD-UHFFFAOYSA-N
Synonym Perfluoroiodocyclohexane
PubChem CID 2759367
IUPAC Name 1,1,2,2,3,3,4,4,5,5,6-undecafluoro-6-iodocyclohexane
SMILES C1(C(C(C(C(C1(F)F)(F)F)(F)I)(F)F)(F)F)(F)F

Specifications

Color Yellow
Boiling Point 112°C
Quantity 1 g
Formula Weight 407.95
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Undecafluoroiodocyclohexane (stabilized with Copper chip)