accessibility menu, dialog, popup

UserName

FFFFFIFFFFFF

CAS RN 355-69-1

IUPAC Name: 1,1,2,2,3,3,4,4,5,5,6-undecafluoro-6-iodocyclohexane
Synonyms: 1,1,2,2,3,3,4,4,5,5,6-undecafluoro-6-iodocyclohexane
Molecular Weight (g/mol): 407.95
Molecular Formula: C6F11I
InChi Key: WYRAAMGCHCLHRD-UHFFFAOYSA-N
SMILES: FC1(F)C(F)(F)C(F)(F)C(F)(I)C(F)(F)C1(F)F
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results