missing translation for 'onlineSavingsMsg'
Learn More

Undecafluorohexanoic Acid High Grade 98.0+%, TCI America™

Catalog No. A57225G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A57225G 5 g
A57221G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A57225G Supplier TCI America Supplier No. A57225G
Only null left

Chemical Identifiers

CAS 307-24-4
Molecular Formula C6HF11O2
Molecular Weight (g/mol) 314.054
MDL Number MFCD00198040
InChI Key PXUULQAPEKKVAH-UHFFFAOYSA-N
Synonym Perfluorohexanoic Acid High Grade, IPC-PFFA-6 HG
PubChem CID 67542
ChEBI CHEBI:83492
IUPAC Name 2,2,3,3,4,4,5,5,6,6,6-undecafluorohexanoic acid
SMILES C(=O)(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)O

Specifications

Melting Point 14°C
Color Colorless
Boiling Point 157°C
Quantity 5 g
UN Number 3265
Formula Weight 314.05
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material Undecafluorohexanoic Acid High Grade [Ion-Pair Reagent for LC-MS]
EINECSNumber (2)-1182
TSCA Yes