CAS RN 307-24-4
IUPAC Name:
undecafluorohexanoic acid
Synonyms:
undecafluorohexanoic acid
Molecular Weight (g/mol):
314.05
Molecular Formula:
C6HF11O2
InChi Key:
PXUULQAPEKKVAH-UHFFFAOYSA-N
SMILES:
OC(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
1
–
2
of
2
results
1
Undecafluorohexanoic Acid (ca. 5mmol) 98.0+%, TCI America™
CAS: 307-24-4 Molecular Formula: C6HF11O2 Molecular Weight (g/mol): 314.054 MDL Number: MFCD00198040 InChI Key: PXUULQAPEKKVAH-UHFFFAOYSA-N Synonym: IPC-PFFA-6, Perfluorohexanoic Acid PubChem CID: 67542 ChEBI: CHEBI:83492 IUPAC Name: 2,2,3,3,4,4,5,5,6,6,6-undecafluorohexanoic acid SMILES: C(=O)(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)O
2
Undecafluorohexanoic Acid 98.0+%, TCI America™
CAS: 307-24-4 Molecular Formula: C6HF11O2 Molecular Weight (g/mol): 314.054 MDL Number: MFCD00198040 InChI Key: PXUULQAPEKKVAH-UHFFFAOYSA-N Synonym: Perfluorohexanoic Acid PubChem CID: 67542 ChEBI: CHEBI:83492 IUPAC Name: 2,2,3,3,4,4,5,5,6,6,6-undecafluorohexanoic acid SMILES: C(=O)(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)O