missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ULYSIS™ Alexa Fluor™ Nucleic Acid Labeling Kit

Catalog No. U21650
Encompass
Change view
Click to view available options
Label or Dye:
Alexa Fluor 488
Alexa Fluor 546
Alexa Fluor 594
Alexa Fluor 647
Excitation/Emission:
492/520 nm
555/570 nm
588/615 nm
650/670 nm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Label or Dye Excitation/Emission
U21650 Alexa Fluor 488 492/520 nm
U21652 Alexa Fluor 546 555/570 nm
U21654 Alexa Fluor 594 588/615 nm
U21660 Alexa Fluor 647 650/670 nm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. U21650 Supplier Invitrogen™ Supplier No. U21650

Achieve unparalleled precision and sensitivity in your fluorescence-based applications from FISH to real-time PCR. With a streamlined, non-enzymatic labeling process and a broad selection of vibrant dyes, ULYSIS kits ensure your nucleic acids are ready for any challenge.

ULYSIS nucleic acid labeling kits provide a unique method for attaching a fluorescent dye to nucleic acids. The labeling reagent in the kit reacts with the N7 of guanine to form a stable coordination complex, and the reaction is simple and fast - just heat denature DNA (5 minutes), add the label (react for 15 minutes), then purify.

Features include:

  • Labeling reaction complete in as little as 15 minutes
  • Available in several Alexa Fluor dye colors
  • Non-enzymatic labeling—avoids the potential biases and limitations associated with enzymatic labeling methods
  • Flexibility—compatible with a wide range of experimental conditions and protocols
  • High sensitivity—provides strong fluorescence signals, enabling detection of low-abundance targets

Reliable labeling with the Universal Linkage System

This series of ULYSIS kits was developed to enable rapid and simple coupling of Alexa Fluor dyes to purine bases in nucleic acid polymers. The method, the Universal Linkage System (ULS), makes use of a platinum dye complex that forms a stable adduct with the N7 position of guanine and, to a lesser extent, adenine bases in DNA, RNA, PNA, and oligonucleotides. The result is a reliable nonenzymatic method for nucleic acid labeling.

Labeling is fast and easy

The labeling reaction typically takes only 15 minutes, and separation of the labeled nucleic acids from the unreacted ULS complex can be accomplished with a simple spin-column procedure. DNA longer than ∼1,000 base pairs require a 10-minute DNase digestion before labeling, which both optimizes labeling and fragments the probe for efficient hybridization.

Specifications

Content And Storage Store in freezer (-5 to -30°C) and protect from light.

Needed but not included: gel filtration-based spin columns for purification of the labeled DNA from excess labeling reagent
Color Green
Detection Method Fluorescence
Format Kit
Label or Dye Alexa Fluor 488
Final Product Type Probes (Labeled RNA), Probes (Labeled DNA), Oligos (Labeled)
Sample Type DNA/RNA
Labeling Target DNA (General), Oligos, RNA (General)
Excitation/Emission 492/520 nm
For Use With (Application) Dot blot, Northern blot, Southern blot, RNA in situ hybridization, DNA in situ hybridization, Multicolor fluorescence in situ hybridization (mFISH), Comparative genome hybridization (CGH), Microarray analysis
Includes Label or Dye Yes
Labeling Method Direct Labeling
Product Line Alexa Fluor, Ulysis
Product Type Nucleic Acid Labeling Kit
Quantity 1 Kit
Shipping Condition Room Temperature
Show More Show Less
Is ULYSIS labeling compatible with microarray analysis?

Yes, there are numerous examples of ULYSIS labeled probes that have been used in microarray analysis. Here are a few publications for your reference:

- Babak T, Zhang W, Marros Q et al. (2004) Probing microRNAs with microarrays: tissue specificity and functional inference. RNA 10(11):1813-1819.
- Hiley SL, Jackman J, Babak T et al. (2005) Detection and discovery of RNA modifications using microarrays. Nucleic Acids Res 33(1):e2.
- Torchet C, Badis G, Devaux F et al. (2005) The complete set of H/ACA snoRNAs that guide rRNA pseudouridylations in Saccharomyces cerevisiae. RNA 11(6):928-938.

Can probes labeled with the ULYSIS Nucleic Acid Labeling Kits be stored for later use?

Long-term storage for the ULYSIS labeled probes can be done in just about any kind of buffer, TE, formamide, hybridization buffer, or ethanol. We suggest using your normal storage conditions as long as you protect the probes from light. ULYSIS conjugates are very stable. Avoid phenol.

Do you have any tips on using the ULYSIS Nucleic Acid Labeling Kits for RNA labeling?

A preliminary protocol modifies our DNA-labeling protocol: Do not nuclease-treat the RNA, but label it directly by incubating for 10 minutes at 90°C or 15 minutes at 85°C. Add 2 µg of glycogen for every 1 µg of RNA and purify by ethanol precipitation. Refer to these publications:

- Babak T, Zhang W, Marros Q et al. (2004) Probing microRNAs with microarrays: tissue specificity and functional inference. RNA 10(11):1813-1819.
- Hiley SL, Jackman J, Babak T et al. (2005) Detection and discovery of RNA modifications using microarrays. Nucleic Acids Res 33(1):e2.
- Torchet C, Badis G, Devaux F et al. (2005) The complete set of H/ACA snoRNAs that guide rRNA pseudouridylations in Saccharomyces cerevisiae. RNA 11(6):928-938.

Can the ULYSIS kits be used on probes longer than 1,000 base pairs or even plasmids?

It might be possible to label larger probes with the ULYSIS Nucleic Acid Labeling Kits, but the dye will likely need to be diluted to avoid (or at least reduce) problems with aggregation. Refer to this publication: Coelho-Castelo AA, Santos Junior RR, Bonato VL et al. (2003) B-lymphocytes in bone marrow or lymph nodes can take up plasmid DNA after intramuscular delivery. Hum Gene Ther 14(13):1279-1285.

How stable is the ULYSIS labeled DNA to high temperature?

An oligonucleotide labeled with a ULYSIS Nucleic Acid Labeling Kit should survive 100°C for 5 minutes, and storage at 68°C overnight should also not cause any dissociation of the complex.

My recovery of DNA after labeling with ULYSIS Nucleic Acid Labeling Kit, and precipitation is low. What can I do to improve it?

After labeling and purifying your DNA, you can add 10 µg of salmon sperm DNA. This should improve your recovery after precipitation.

Can I use the DNA labeled using ULYSIS Nucleic Acid Labeling Kit, for hybridization directly after column purification?

We recommend using ethanol precipitation to concentrate the sample first to calculate the DNA concentration and degree of labeling.

Could you provide an overview of the ULYSIS Nucleic Acid Labeling Kits?

The ULYSIS oligonucleotide chemical labeling reagents allow you to rapidly and easily couple our fluorescent dyes to purine bases in nucleic acid polymers. The method, the Universal Linkage System (ULS), is based on the use of a platinum coordination complex that forms a stable adduct at the N7 position of guanine. ULYSIS labeled probes are used in fluorescence in situ hybridization (FISH) and numerous other applications.

What are the ULYSIS labeling kits used for?

ULYSIS labeling kits are used for labeling short, unmodified DNA and RNA strands between ~100-1,000 bp for FISH analysis.


For Research Use Only. Not for use in diagnostic procedures.