missing translation for 'onlineSavingsMsg'
Learn More

Ultramark∣r 1621, Mass Spec Std

Catalog No. aal16698md
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL16698MD 250 mg
AAL1669803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL16698MD Supplier Thermo Scientific Chemicals Supplier No. L16698MD
Only null left

CAS: 105809-15-2 | C6H18N3P3 | 231.189 g/mol

Ultramark 1621 is used as a reference compound for positive and negative ion fast-atom bombardment high-resolution mass spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ultramark 1621 is used as a reference compound for positive and negative ion fast-atom bombardment high-resolution mass spectrometry.

Solubility
Miscible with water.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 105809-15-2
Molecular Formula C6H18N3P3
Molecular Weight (g/mol) 231.189
MDL Number MFCD00242454
InChI Key DEBZEVJNWCNATM-MZWXYZOWSA-N
PubChem CID 44119860
IUPAC Name 2,2,4,4,6,6-hexakis(deuteriomethyl)-1,3,5-triaza-2$l^{5},4$l^{5},6$l^{5}-triphosphacyclohexa-1,3,5-triene
SMILES CP1(=NP(=NP(=N1)(C)C)(C)C)C

Specifications

Density 1.8
Solubility Miscible with water
Quantity 250 mg
Formula Weight 2120
Chemical Name or Material Ultramark™ 1621, Mass Spec Standard
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only