missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Wearwell™ Ultra Tred ArmorCote Mat
Urethane coating gives this antifatigue mat extra durability
$193.66 - $503.56
Specifications
| For Use With (Application) | Labs, pharmacies, hospitals, industrial settings |
|---|
Description
- PVC sponge antifatigue mat with a urethane wear-layer for longer performance
- The double-thick urethane coating offers more abrasion and chemical resistance than comparable sponge products
- Ideal for use in mostly dry applications such as labs and other medium traffic areas
- 0.375 in. thick with eye-catching yellow safety borders
- Color: Black with yellow border
Order Info
Available in cut lengths up to 60 ft. (18m) in 3 ft. widths (0.9m). Contact your Fisher Safety Customer Service Representative for details.
Specifications
| Labs, pharmacies, hospitals, industrial settings |