missing translation for 'onlineSavingsMsg'
Learn More

Wearwell™ Ultra Tred ArmorCote Mat

Catalog No. 17977503
Change view
Click to view available options
Dimensions (L x W):
3 x 5 ft.
3 x 12 ft.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
17977503 3 x 12 ft.
17986123 3 x 5 ft.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17977503 Supplier Wearwell™ Supplier No. 440.38X3X12BYL

Urethane coating gives this antifatigue mat extra durability

  • PVC sponge antifatigue mat with a urethane wear-layer for longer performance
  • The double-thick urethane coating offers more abrasion and chemical resistance than comparable sponge products
  • Ideal for use in mostly dry applications such as labs and other medium traffic areas
  • 0.375 in. thick with eye-catching yellow safety borders
  • Color: Black with yellow border

Order Info

Available in cut lengths up to 60 ft. (18m) in 3 ft. widths (0.9m). Contact your Fisher Safety Customer Service Representative for details.

Specifications

Dimensions (L x W) 3 x 12 ft.
For Use With (Application) Labs, pharmacies, hospitals, industrial settings