missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Ultra Streptavidin-HRP

Catalog No. FERN504
Change view
Click to view available options
Quantity:
1.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERN504 1.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FERN504 Supplier Thermo Scientific™ Supplier No. N504

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Ultra Streptavidin-HRP

Streptavidin-HRP consists of streptavidin protein that is covalently conjugated to horseradish peroxidase (HRP) enzyme. Streptavidin binds to biotin and the conjugated HRP provides a high level of activity for detection using an appropriate substrate system.

Features of Streptavidin-HRP:

  • Product Number: N504
  • Size: 1.5 mL
  • Formulation: Conjugate in stabilizer solution containing protein and preserved with 0.02% methyliso-thiazolone, 0.02% bromonitrodioxane, and 0.002% other active isothiazolones.
  • Storage: Shipped on cold packs. Store at 2-8°C. Stable until expiration date if stored under these conditions.
  • Application: ELISA: to be used with biotin-labeled detection antibodies. Recommended dilution range is 1:400 - 1:2,000 in ELISA diluent (e.g., PBS containing 4% protease-free BSA) . The optimal dilution must be determined empirically for each specific application. Prepare dilutions immediately before use.
  • Specifications

    Conjugate HRP
    Quantity 1.5 mL
    Form Liquid
    Product Type Streptavidin Conjugate
    Target Molecule Biotin
    Label or Dye HRP (Horseradish Peroxidase)

    For Research Use Only. Not for use in diagnostic procedures.