Learn More
Greiner Bio-One Ultra Low Profile Polystyrene Microplate Lids
Used for high-format low profile microplates
$923.93 - $1004.89
Specifications
| Color | Clear |
|---|---|
| Material | Polystyrene |
| Condensation Rings | No |
| No. per Case | 100 |
| No. per Pack | 5 |
Description
- Free of detectable DNase, RNase, human DNA
- Non-Pyrogenic, non-cytotoxic
- Offer protection against contamination and evaporation during sample storage and cell cultivation
- Compatible with a range of different microplates
Specifications
| Clear | |
| No | |
| 5 |
| Polystyrene | |
| 100 | |
| A variety of microplates, particularly well suited for high-format, low-profile microplates |
For Research Use Only. Not for use in diagnostic procedures, unless otherwise indicated.