missing translation for 'onlineSavingsMsg'
Learn More

PIG™ Ultra-Duty Adhesive-Backed Heavyweight Grippy Roll

Heavyweight adhesive-backed ultra-duty floor mat creates a safe walking surface in entrances and aisleways.

Supplier:  PIG™ GRP36301GY

 View more versions of this product

Catalog No. 18366540


Only null left

Description

Description

  • Provides a high traction surface
  • Adhesive-backed floor mat stays exactly where you put it with no shifting, bunching up or ripples
  • Dries out quickly, and liquid-proof barrier layer prevents moisture from soaking through to the floor
  • Absorbs water, oil and grease and traps dirt and salt to keep it from being tracked around
  • Stays in place for up to 3 to 6 months, depending on conditions and traffic patterns
  • Easy to clean
  • Tested and Certified by the National Floor Safety Institute
  • For indoor use on concrete, vinyl, linoleum, ceramic tile, quarry tile and laminate flooring
  • 1 - 3 x 50 ft. roll
Specifications

Chemical Identifiers

50 ft.
Plastic
3 ft.
15.2 m
Adhesive Floor Mat
91 cm

Specifications

Smoke Gray
15.2 m
91 cm
3 ft.
50 ft.
Plastic
Adhesive Floor Mat
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions