missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Azure Biosystems Ultra Clear Flat 8-Cap Strip
Description
- Cap strips designed to fit on most tube strips, and 96-well plates. The specially designed indented cap prevents 'finger touch' signal interference.
- Flat-top profile and suitable for qPCR assays. They are available in 8-cap strips and provide a tight seal.
- Ultra clear
- Optical wide area 8-cap strip
- Pack of 120
Specifications
Specifications
| Product Type | Microtube Cap |
| Autoclavable | Not Autoclavable |
| Material | Ethylene Propylene |
| Color | Clear |
| Sterility | Non-sterile |
| For Use With (Equipment) | Real-time PCR |
| Disposable | Single-use |