missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ULK2 Monoclonal Antibody (2H4B2)
GREENER_CHOICE

Catalog No. PIMA531948
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA531948 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA531948 Supplier Invitrogen™ Supplier No. MA531948
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Mouse Monoclonal Antibody

MA5-31948 has been tested in indirect ELISA.

ULK2, also known as ATG1B, is a key serine/threonine protein kinase probably acting at the most upstream step of autophagosome formation. Knockout of ULK2 results in a severe defect in the autophagy pathway. ULK2 is highly conserved among eukaryotes and shows high homology with its related protein ULK1. Both ULK1 and ULK2 form a complex with ATG13 and FIP200 that mediates TOR signaling and is essential for autophagy. Like ULK1, ULK2 is also thought to be involved in early neuronal growth and differentiation.

Specifications

Antigen ULK2
Applications ELISA, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 2H4B2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene ULK2
Gene Accession No. Q8IYT8, Q9QY01
Gene Alias A830085I22Rik; ATG1B; AU015340; EC:2.7.11.1; KIAA0623; mKIAA0623; serine/threonine-protein kinase ULK2; Serine/threonine-protein kinase Unc51.2; ULK2; unc-51 like autophagy activating kinase 2; unc-51 like kinase 2; Unc51.2; unc-51-like kinase 2
Gene Symbols ULK2
Host Species Mouse
Immunogen Purified recombinant fragment of human ULK2 (AA: 1-155) expressed in E. coli.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29869, 303206, 9706
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less