missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ulipristal Acetate 98.0+%, TCI America™
$307.57 - $1074.62
Chemical Identifiers
| CAS | 126784-99-4 |
|---|---|
| Molecular Formula | C30H37NO4 |
| Molecular Weight (g/mol) | 475.629 |
| MDL Number | MFCD00899035 |
| InChI Key | OOLLAFOLCSJHRE-ZHAKMVSLSA-N |
| Synonym | (11beta)-17-(Acetyloxy)-11-[4-(dimethylamino)phenyl]-19-norpregna-4,9-diene-3,20-dione, CDB-2914 |
| PubChem CID | 130904 |
| ChEBI | CHEBI:71025 |
| IUPAC Name | [(8S,11R,13S,14S,17R)-17-acetyl-11-[4-(dimethylamino)phenyl]-13-methyl-3-oxo-1,2,6,7,8,11,12,14,15,16-decahydrocyclopenta[a]phenanthren-17-yl] acetate |
| SMILES | CC(=O)C1(CCC2C1(CC(C3=C4CCC(=O)C=C4CCC23)C5=CC=C(C=C5)N(C)C)C)OC(=O)C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
U010250MG
|
TCI America
U010250MG |
50 mg |
Each for $307.57
|
|
|||||
|
U0102250MG
|
TCI America
U0102250MG |
250 mg |
Each for $1,074.62
|
|
|||||
Chemical Identifiers
| 126784-99-4 | |
| 475.629 | |
| OOLLAFOLCSJHRE-ZHAKMVSLSA-N | |
| 130904 | |
| [(8S,11R,13S,14S,17R)-17-acetyl-11-[4-(dimethylamino)phenyl]-13-methyl-3-oxo-1,2,6,7,8,11,12,14,15,16-decahydrocyclopenta[a]phenanthren-17-yl] acetate |
| C30H37NO4 | |
| MFCD00899035 | |
| (11beta)-17-(Acetyloxy)-11-[4-(dimethylamino)phenyl]-19-norpregna-4,9-diene-3,20-dione, CDB-2914 | |
| CHEBI:71025 | |
| CC(=O)C1(CCC2C1(CC(C3=C4CCC(=O)C=C4CCC23)C5=CC=C(C=C5)N(C)C)C)OC(=O)C |
Specifications
| 126784-99-4 | |
| Yellow | |
| MFCD00899035 | |
| (11beta)-17-(Acetyloxy)-11-[4-(dimethylamino)phenyl]-19-norpregna-4,9-diene-3,20-dione, CDB-2914 | |
| CC(=O)C1(CCC2C1(CC(C3=C4CCC(=O)C=C4CCC23)C5=CC=C(C=C5)N(C)C)C)OC(=O)C | |
| 475.629 | |
| CHEBI:71025 | |
| ≥98.0% (HPLC,T) | |
| Ulipristal Acetate |
| 186°C | |
| C30H37NO4 | |
| 50 mg | |
| OOLLAFOLCSJHRE-ZHAKMVSLSA-N | |
| [(8S,11R,13S,14S,17R)-17-acetyl-11-[4-(dimethylamino)phenyl]-13-methyl-3-oxo-1,2,6,7,8,11,12,14,15,16-decahydrocyclopenta[a]phenanthren-17-yl] acetate | |
| 130904 | |
| 475.63 | |
| Crystalline Powder |
Safety and Handling
TSCA : No
Recommended Storage : Freezer (-20 °C)