missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ulipristal Acetate 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 186°C |
| Color | Yellow |
| Quantity | 50 mg |
| Formula Weight | 475.63 |
| Percent Purity | ≥98.0% (HPLC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Ulipristal Acetate |
Safety and Handling
| TSCA | No |
| Recommended Storage | Freezer (-20 °C) |