missing translation for 'onlineSavingsMsg'
Learn More

Cupric Acetate, Monohydrate, Crystal, ACS, 98-102%, Spectrum™ Chemical

Catalog No. 18609970
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18609970 50 kg Poly Bottle
18609966 125 g Poly Bottle
18609967 500 g Poly Bottle
18609968 2.5 kg Poly Bottle
18609969 12 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18609970 Supplier Spectrum Chemical Mfg Cor Supplier No. C137050KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6046-93-1 | 199.65 g/mol

Chemical Identifiers

CAS 6046-93-1

Specifications

CAS Max % 1
Physical Form Crystals or Needles
Quantity 50 kg
Molecular Weight (g/mol) 199.65
Percent Purity 98 to 102%
Grade ACS/Reagent
Packaging Poly Bottle
Chemical Name or Material Cupric Acetate, Monohydrate, Crystal

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.