missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ UFC1 Recombinant Rabbit Monoclonal Antibody (23GB4245)

Catalog No. MA553717
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA553717 20 μL
MA553716 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA553717 Supplier Invitrogen™ Supplier No. MA553717
Only null left

Rabbit Recombinant Monoclonal Antibody

UFC1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow

UFC1 is an E2-like conjugating enzyme for ubiquitin-fold modifier-1 (Komatsu et al., 2004 ).

Specifications

Antigen UFC1
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB4245
Concentration 2.0 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene UFC1
Gene Accession No. Q9Y3C8
Gene Alias 1110021H02Rik; AA062310; CGI-126; ESTM29; HSPC155; ubiquitin-fold modifier conjugating enzyme 1; ubiquitin-fold modifier-conjugating enzyme 1; UFC1; ufc1-s; Ufm1-conjugating enzyme 1; Ufm1-conjugating enzyme 1-
Gene Symbols UFC1
Host Species Rabbit
Immunogen A synthesized peptide derived from UFC1 (1-130AA).
Purification Method Affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51506
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less