missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ UDP-alpha-D-Galactose, Disodium Salt, >95% Calbiochem™,

Catalog No. 67011150mg
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
67011150MG 50 mg
67011110MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 67011150MG Supplier MilliporeSigma™ Supplier No. 67011150MG

A donor substrate for galactosyltransferases used in the biosynthesis of galactose-containing oligosaccharides

Chemical Identifiers

CAS 137868-52-1
Molecular Formula C15H22N2Na2O17P2
Molecular Weight (g/mol) 610.27
MDL Number MFCD00077895
InChI Key PKJQEQVCYGYYMM-UHFFFAOYNA-L
Synonym udp-a-d-galactose disodium salt
PubChem CID 126969254
IUPAC Name disodium [5-(2,4-dioxo-1,2,3,4-tetrahydropyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl {[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl phosphonato]oxy}phosphonate
SMILES [Na+].[Na+].OCC1OC(OP([O-])(=O)OP([O-])(=O)OCC2OC(C(O)C2O)N2C=CC(=O)NC2=O)C(O)C(O)C1O

Specifications

Color White
Quantity 50 mg
Solubility Information Easily soluble in cold water
Formula Weight 610.3
Percent Purity ≥95%
Grade HPLC
Physical Form Solid
Chemical Name or Material UDP-alpha-D-Galactose, Disodium Salt
Recommended Storage Keep container tightly closed. Keep container in a cool, well-ventilated area. Do not store above -20°C (-4°F).