CAS RN 137868-52-1
IUPAC Name:
disodium [5-(2,4-dioxo-1,2,3,4-tetrahydropyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl {[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl phosphonato]oxy}phosphonate
Synonyms:
disodium [5-(2,4-dioxo-3H-pyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl [3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl phosphonato]oxyphosphonate
Molecular Weight (g/mol):
610.27
Molecular Formula:
C15H22N2Na2O17P2
InChi Key:
PKJQEQVCYGYYMM-UHFFFAOYNA-L
SMILES:
[Na+].[Na+].OCC1OC(OP([O-])(=O)OP([O-])(=O)OCC2OC(C(O)C2O)N2C=CC(=O)NC2=O)C(O)C(O)C1O
1
–
1
of
1
results
1
MilliporeSigma™ UDP-alpha-D-Galactose, Disodium Salt, >95% Calbiochem™,
CAS: 137868-52-1 Molecular Formula: C15H22N2Na2O17P2 Molecular Weight (g/mol): 610.27 MDL Number: MFCD00077895 InChI Key: PKJQEQVCYGYYMM-UHFFFAOYNA-L Synonym: udp-a-d-galactose disodium salt PubChem CID: 126969254 IUPAC Name: disodium [5-(2,4-dioxo-1,2,3,4-tetrahydropyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl {[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl phosphonato]oxy}phosphonate SMILES: [Na+].[Na+].OCC1OC(OP([O-])(=O)OP([O-])(=O)OCC2OC(C(O)C2O)N2C=CC(=O)NC2=O)C(O)C(O)C1O