Learn More
Invitrogen™ Human Ubiquitin ELISA Kit
ELISA
$892.00 - $892.00
Specifications
| Accession Number | P62979 |
|---|---|
| Assay Range | 27.43 to 20,000 pg/mL |
| Assay Sensitivity | 30 pg/mL |
| Assay | Solid-phase sandwich ELISA |
| Conjugate | HRP |
Description
The Human Ubiquitin 1 RPS27A ELISA quantitates Hu Ubiquitin in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu Ubiquitin. Principle of the method The Human Ubiquitin solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Ubiquitin, a highly conserved protein that has a major role in targeting cellular proteins for degradation by the 26S proteosome, is synthesized as a precursor protein consisting of either polyubiquitin chains or a single ubiquitin fused to an unrelated protein. This gene encodes a fusion protein consisting of ubiquitin at the N terminus and ribosomal protein S27a at the C terminus. When expressed in yeast, the protein is post-translationally processed, generating free ubiquitin monomer and ribosomal protein S27a. Ribosomal protein S27a is a component of the 40S subunit of the ribosome and belongs to the S27AE family of ribosomal proteins. It contains C4-type zinc finger domains and is located in the cytoplasm. Pseudogenes derived from this gene are present in the genome. As with ribosomal protein S27a, ribosomal protein L40 is also synthesized as a fusion protein with ubiquitin; similarly, ribosomal protein S30 is synthesized as a fusion protein with the ubiquitin-like protein fubi. Multiple alternatively spliced transcript variants that encode the same proteins have been identified.Specifications
| P62979 | |
| 30 pg/mL | |
| HRP | |
| Other Proteins | |
| This ELISA kit shows no cross-reactivity with the following cytokines tested: human Angiogenin, BDNF, BLC, ENA-78, FGF-4, IL-1 A, IL-1 b, IL-2, IL-3, IL-4, IL-5, IL-6, IL-7, IL-8, IL-9, IL-11, IL-12 p70, IL-12 p40, IL-13, IL-15, IL-309, IP-10, G-CSF, GM-CSF, IFN-g, Leptin (OB), MCP-1, MCP-2, MCP-3, MDC, MIP-1 A, MIP-1 b, MIP-1 delta, PARC, PDGF, RANTES, SCF, TARC, TGF-b, TIMP-1, TIMP-2, TNF-a, TNF-b, TPO, VEGF. | |
| ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis | |
| 51065, 6233, 7311, 7314, 7316 | |
| <12% | |
| Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
| RUO | |
| Plasma 50 μL, Serum 50 μL, Supernatant 100 μL | |
| CEP80, HEL112, S27A, UBA80, UBC, UBCEP1, UBCEP80 | |
| 1 hrs 20 mins |
| 27.43 to 20,000 pg/mL | |
| Solid-phase sandwich ELISA | |
| ELISA | |
| Plasma, Serum, Supernatant | |
| Human | |
| Colorimetric Microplate Reader | |
| CEP80,HEL112,S27A,UBA80,UBC,UBCEP1,UBCEP80 | |
| <10 | |
| HRP | |
| Immunology | |
| 2°C to 8°C | |
| Ubiquitin | |
| 4 hrs 45 mins |