missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBE4B Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Description
The recommended shelf life for this product is 1 year from date of receipt.
The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes, or E1s, ubiquitin-conjugating enzymes, or E2s, and ubiquitin-protein ligases, or E3s. UBE4B represents an additional conjugation factor, E4, which is involved in multiubiquitin chain assembly. UBE4B also the strongest candidate among the neuroblastoma tumor suppressors.
Specifications
Specifications
| Antigen | UBE4B |
| Applications | Immunoprecipitation |
| Classification | Polyclonal |
| Concentration | 1 mg/ml |
| Conjugate | Unconjugated |
| Formulation | phosphate, tris citrate with 0.09% sodium azide; pH 7-8 |
| Gene | UBE4B |
| Gene Accession No. | O95155 |
| Gene Alias | Ubiquitin conjugation factor E4 B; Ubiquitin-fusion degradation protein 2; Homozygously deleted in neuroblastoma-1; HDNB1; KIAA0684; UFD2 |
| Gene Symbols | UBE4B |
| Show More |