missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ UBALD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564288
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564288 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564288 Supplier Invitrogen™ Supplier No. PA564288
Only null left

Rabbit Polyclonal Antibody

Immunogen sequence: MFSRLKASES FHSGGSGSPM AATATSPPPH FPHAATSSSA ASSWPTAASP PGGPQHHQPQ PPLWTPTPPS PASDWPPLAP QQATSEPRAH PAMEAER Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 89%.

Specifications

Antigen UBALD1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene UBALD1
Gene Accession No. Q8TB05
Gene Alias FAM100A; family with sequence similarity 100, member A; PP11303; protein FAM100A; UBA like domain containing 1; UBALD1; UBA-like domain containing 1; UBA-like domain-containing protein 1
Gene Symbols UBALD1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human UBALD1. Recombinant protein control fragment (Product #RP-103638).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 124402
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less