missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals U0126, 99+%

Catalog No. AAJ61246MC
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ61246MC 100 mg
AAJ61246MB 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ61246MC Supplier Thermo Scientific Chemicals Supplier No. J61246MC

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Selective inhibitor of mitogen-activated protein kinases MEK-1 and MEK-2 | CAS: 109511-58-2 | C18H16N6S2, C18H16N6S2 | 380.49 g/mol

U0126, is used as a selective MAP Kinase Kinase (MKK, MEK) inhibitor. U-0126 has been observed to inhibit promoters containing an AP-1 response element, while in contrast having no effect on promoters that lack an AP-1 response group. U-0126 is also useful in neuronal studies in mice since it displays inhibitory effects against oxidative stress. U-0126 is an activator of PGC-1, mtTFA and nuclear respiratory factor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 109511-58-2
Molecular Formula C18H16N6S2
Synonym 1,4-Diamino-2,3-dicyano-1,4-bis(2-aminophenylthio)butadiene

Specifications

Quantity 100 mg
Description White
Content And Storage -30°C to -10°C
For Use With (Application) Selective inhibitor of mitogen-activated protein kinases MEK-1 and MEK-2
Formula Weight 380.49g/mol
Solubility Soluble in DMSO; Poorly soluble in ethanol and water
Product Type Signal Transduction Reagent
Form Powder
Assay ≥99%

RUO – Research Use Only