missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals U-73122, 95%

Supplier:  Thermo Scientific Chemicals J62898MCR

 View more versions of this product

Catalog No. AAJ62898MCR


Only null left

Description

Description

It is used as a phospholipase C, phospholipase A2, and 5-LO inhibitor. It is also determined that in SK-N-SH neuroblastoma cells, U-73122 inhibits agonist-induced down-regulation of muscarinic receptors. In addition, it is a useful tool to investigate receptor-mediated PI turnover in signal transduction. U-73122 is a potent inhibitor of human neutrophil adhesion to biological surfaces (IC50 = 50 nM) as well as adhesion-dependent granule exocytosis and oxidative burst. U-73343 (sc-201422) is useful as a negative control for investigations of U-73122 phospholipase C antagonism and its cellular consequences.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Inhibitor of phospholipase C activation and inhibits the down regulation of muscarinic receptors.
Specifications

Specifications

1 mg
Inhibitor of phospholipase C activation and inhibits the down regulation of muscarinic receptors
112648-68-7
464.65g/mol
Soluble in DMSO, ethanol, or DMF at approx. 0.5mg/ml; May require heating or overnight mixing; Insoluble in water
Crystalline solid
COC1=CC=C2[C@H]3CC[C@]4(C)[C@H](CC[C@H]4[C@@H]3CCC2=C1)NCCCCCCN1C(=O)C=CC1=O
464.65
Off-white
≥99%
C29H40N2O3
1-[6-[((17beta)-3-Methoxyestra-1,3,5[10]-trien-17-yl)amino]hexyl]pyrrole-2,5-dione
Signal Transduction Reagent
LUFAORPFSVMJIW-ZRJUGLEFSA-N
1-(6-{[(1S,3aS,3bR,9bS,11aS)-7-methoxy-11a-methyl-1H,2H,3H,3aH,3bH,4H,5H,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-yl]amino}hexyl)-2,5-dihydro-1H-pyrrole-2,5-dione

Chemical Identifiers

112648-68-7
C29H40N2O3
1-[6-[((17beta)-3-Methoxyestra-1,3,5[10]-trien-17-yl)amino]hexyl]pyrrole-2,5-dione
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only