Learn More
DuPont™ Tyvek™ IsoClean™ 255 Series Coveralls
Sterile coverall with collar for use in ISO 6 (Class 1,000) or higher controlled environments.
Supplier:  DuPont™ IC255BWH5X0025CS
 View more versions of this product
Catalog No. 18-888-117
Only null left
Explore available promotions
Description
Description
  • Tyvek flashspun polyolefin protective material
  • Patented flash-spinning process
  • Creates barrier to dry particles, microorganisms, non-hazardous liquids
  • Lightweight, durable
  • Collared
  • Elastic thumb loops keep sleeves in place
Specifications
Specifications
Cleanroom Coveralls
Unisex
White
Covered Elastic
Zipper
Sterile
IC255BWH
Protective Barrier to Dry Particle, Microorganisms
5X-Large
Tyvek
Single-use
Standard
Bound
CE
Set sleeves
Product Suggestions
Product Suggestions