missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ Tyvek™ IsoClean™ 255 Series Coveralls
GREENER_CHOICE

Catalog No. p-7079215
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
6X-Large
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
18888110 Small
18888111 Medium
18888112 Large
18888113 X-Large
18888114 2X-Large
18888115 3X-Large
18888116 4X-Large
18888117 5X-Large
18888118 6X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 18888110 Supplier DuPont™ Supplier No. IC255BWHSM0025CS
Only null left

Sterile coverall with collar for use in ISO 6 (Class 1,000) or higher controlled environments.

  • Tyvek flashspun polyolefin protective material
  • Patented flash-spinning process
  • Creates barrier to dry particles, microorganisms, non-hazardous liquids
  • Lightweight, durable
  • Collared
  • Elastic thumb loops keep sleeves in place

Specifications

Product Type Cleanroom Coveralls
Size Small
Gender Unisex
Material Tyvek
Color White
Disposable Single-use
Ankle Style Covered Elastic
Hood Type Standard
Closure Type Zipper
Seam Type Bound
Sterility Sterile
Certifications/Compliance (Europe) CE
Model IC255BWH
Sleeve Style Set sleeves
For Use With (Application) Protective Barrier to Dry Particle, Microorganisms
Show More Show Less