missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ Tyvek™ IsoClean™ 255 Series Coveralls
Sterile coverall with collar for use in ISO 6 (Class 1,000) or higher controlled environments.
Supplier: DuPont™ IC255BWH2X0025CS
Description
- Tyvek flashspun polyolefin protective material
- Patented flash-spinning process
- Creates barrier to dry particles, microorganisms, non-hazardous liquids
- Lightweight, durable
- Collared
- Elastic thumb loops keep sleeves in place
Specifications
| Cleanroom Coveralls | |
| Unisex | |
| White | |
| Covered Elastic | |
| Zipper | |
| Sterile | |
| IC255BWH | |
| Protective Barrier to Dry Particle, Microorganisms |
| 2X-Large | |
| Tyvek | |
| Single-use | |
| Standard | |
| Bound | |
| CE | |
| Set sleeves |