missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ Tyvek™ IsoClean™ Coverall (all)
Tyvek non-sterile cleanroom coveralls protect workers from non-hazardous dry particulates and liquid splash.
Supplier: DuPont™ IC181SWHXL002500
Alert:
Garments made of Tyvek should not be used around heat, flames, sparks or in potentially flammable or explosive environments.
Description
- Serged seams have multiple interlocking threads that are sewn around the raw edges of garment material to create a strong, stress-resistant seam.
- Coverall has elastic openings for tighter fit at wrist and ankle.
- Laydown collar
- Front zipper closure for easy donning and doffing.
- Bulk packaged in double transparent poly liners.
- Pin locking slider locks in place when the pins on the puller are pushed into the zipper elements
- Manufactured under specifications that do not contain natural rubber latex
Specifications
| Cleanroom Coveralls | |
| Unisex | |
| White | |
| Elastic | |
| Zipper | |
| Non-sterile | |
| Elastic wrist and ankle, standard collar with set sleeves | |
| 29.5 in. | |
| 74.9 cm | |
| Barrier to Dry Particles, Microorganisms, and Non-Hazardous Liquids in Controlled Environments | |
| 92.71 cm | |
| 5.11 to 6.3 ft. | |
| Class 1000 (ISO 6) |
| X-Large | |
| Tyvek | |
| Single-use | |
| Elastic | |
| Serged | |
| Standard fold | |
| 40 to 43 in. | |
| 101.6 to 109.22 cm | |
| 5.4 mil | |
| 36.5 in. | |
| 27.25 in. | |
| 155.75 to 192.04 cm |