missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ Tyvek™ IsoClean™ Coverall (all)
GREENER_CHOICE

Catalog No. p-3836414
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19100403 Small
19042440 Medium
17988020 2X-Large
17988018 Large
17988021 3X-Large
17988022 4X-Large
17988019 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Garments made of Tyvek should not be used around heat, flames, sparks or in potentially flammable or explosive environments.
Catalog No. 19100403 Supplier DuPont™ Supplier No. IC181SWHSM002500
Only null left
Garments made of Tyvek should not be used around heat, flames, sparks or in potentially flammable or explosive environments.

Tyvek non-sterile cleanroom coveralls protect workers from non-hazardous dry particulates and liquid splash.

  • Serged seams have multiple interlocking threads that are sewn around the raw edges of garment material to create a strong, stress-resistant seam.
  • Coverall has elastic openings for tighter fit at wrist and ankle.
  • Laydown collar
  • Front zipper closure for easy donning and doffing.
  • Bulk packaged in double transparent poly liners.
  • Pin locking slider locks in place when the pins on the puller are pushed into the zipper elements
  • Manufactured under specifications that do not contain natural rubber latex

Specifications

Product Type Cleanroom Coveralls
Size Small
Gender Unisex
Material Tyvek
Color White
Disposable Single-use
Ankle Style Elastic
Cuff Style Elastic
Closure Type Zipper
Seam Type Serged
Sterility Non-sterile
Collar Type Standard fold
Description Elastic wrist and ankle, standard collar with set sleeves
Size (English) Chest 32 to 36 in.
Size (English) Inseam 27.5 in.
Size (Metric) Chest 81.28 to 91.44 cm
Size (Metric) Inseam 69.85 cm
Thickness 5.4 mil
For Use With (Application) Barrier to Dry Particles, Microorganisms, and Non-Hazardous Liquids in Controlled Environments
Length (English) Sleeve 32.5 in.
Length (Metric) Sleeve 82.55 cm
Height (English) 5 to 5.7 ft.
Height (Metric) 152.4 to 173.73 cm
Cleanroom Class Class 1000 (ISO 6)
Show More Show Less