Learn More
DuPont™ Tyvek™ 500 HV Coverall w/ Elastic Wrists and Ankles
500 HV garments are mainly composed of flash spun polyethylene, providing an ideal balance of protection, durability and comfort
Supplier:  DuPont™ TY125SHV4X0025XC
 View more versions of this product
Catalog No. 17000532
Only null left
Explore available promotions
Description
Description
  • EN 14126 (barrier to infective agents), EN 1073-2 (protection against radioactive contamination), EN ISO 20471 (High Visibility Clothing, Class 3)
  • Antistatic treatment (EN 1149-5) - on inside
  • Stitched external seams
  • Very low inward leakage thanks to optimized design
  • Tyvek™ auto-lock zipper and zipper flap for increased protection.
  • Pin locking slider locks in place when the pins on the puller are pushed into the zipper elements
  • Collared coverall. Mandarin collar
  • Products made after January, 2023 are manufactured under specifications that do not contain natural rubber latex
Specifications
Specifications
High Visibility Coveralls
Unisex
Fluorescent Orange, Orange
Elastic
Elastic
Serged
4X-Large
Tyvek
Single-use
Elastic
Zipper
EN 14126 (barrier to infective agents), EN 1073-2 (protection against radioactive contamination), EN ISO 20471 (High Visibility Clothing, Class 3), Antistatic treatment (EN 1149-5) on inside
Product Suggestions
Product Suggestions