Learn More
DuPont™ Tyvek™ 500 HV Coverall w/ Elastic Wrists and Ankles
500 HV garments are mainly composed of flash spun polyethylene, providing an ideal balance of protection, durability and comfort
Supplier:  DuPont™ TY125SHV2X0025XC
 View more versions of this product
Catalog No. 17000530
Only null left
Explore available promotions
Description
Description
  • EN 14126 (barrier to infective agents), EN 1073-2 (protection against radioactive contamination), EN ISO 20471 (High Visibility Clothing, Class 3)
  • Antistatic treatment (EN 1149-5) - on inside
  • Stitched external seams
  • Very low inward leakage thanks to optimized design
  • Tyvek™ auto-lock zipper and zipper flap for increased protection.
  • Pin locking slider locks in place when the pins on the puller are pushed into the zipper elements
  • Collared coverall. Mandarin collar
  • Products made after January, 2023 are manufactured under specifications that do not contain natural rubber latex
Specifications
Specifications
High Visibility Coveralls
Unisex
Fluorescent Orange, Orange
Elastic
Elastic
Serged
EN 14126 (barrier to infective agents), EN 1073-2 (protection against radioactive contamination), EN ISO 20471 (High Visibility Clothing, Class 3), Antistatic treatment (EN 1149-5) on inside
2X-Large
Tyvek
Single-use
Elastic
Zipper
ANSI/ISEA 107-2020 Type R/Class 3 Section 11
Product Suggestions
Product Suggestions