Learn More
DuPont™ Tyvek™ 500 HV Coverall w/ Elastic Wrists and Ankles
500 HV garments are mainly composed of flash spun polyethylene, providing an ideal balance of protection, durability and comfort
Supplier: DuPont™ TY125SHVMD0025XC
Description
- EN 14126 (barrier to infective agents), EN 1073-2 (protection against radioactive contamination), EN ISO 20471 (High Visibility Clothing, Class 3)
- Antistatic treatment (EN 1149-5) - on inside
- Stitched external seams
- Very low inward leakage thanks to optimized design
- Tyvek™ auto-lock zipper and zipper flap for increased protection.
- Pin locking slider locks in place when the pins on the puller are pushed into the zipper elements
- Collared coverall. Mandarin collar
- Products made after January, 2023 are manufactured under specifications that do not contain natural rubber latex
Specifications
| High Visibility Coveralls | |
| Unisex | |
| Fluorescent Orange, Orange | |
| Elastic | |
| Elastic | |
| Serged |
| Medium | |
| Tyvek | |
| Single-use | |
| Elastic | |
| Zipper | |
| EN 14126 (barrier to infective agents), EN 1073-2 (protection against radioactive contamination), EN ISO 20471 (High Visibility Clothing, Class 3), Antistatic treatment (EN 1149-5) on inside |