Learn More
DuPont™ Tyvek™ 500 Hooded Coverall
DuPont™ Tyvek™ 500 coveralls offer enhanced protection againt a range of threats, from waterbased chemical liquid splashes, asbestos, pharmaceutical and radioactive particulates to biological hazards.
$449.40 - $449.40
Specifications
Gender Unisex
Material Tyvek
Color White
Disposable Single-use
Waist Style Elastic (Glued-in)
View More Specs
Products 1
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
17-500-257
View Documents
DuPont™
TY198SWH6X0025LA
6X-Large N/A
Only null left
Description
Description
  • Pin locking slider locks in place when the pins on the puller are pushed into the zipper elements
  • Products made after January, 2023 are manufactured under specifications that do not contain natural rubber latex.
  • Protection against infective agents (EN 14126) including resistance to penetration by blood and body fluids using synthetic blood (ISO 16603 Class 3) and resistance to penetration by blood-borne pathogens using Phi-X174 bacteriophage (ISO 16604 Class 2)
  • Chemical protective clothing, Category III, Type 5-B and 6-B. (1956)
  • EN 1073-2 (protection against radioactive contamination)
  • Antistatic treatment (EN 1149-5) - on both sides.
  • Serged external seams featuring patent-pending technology for enhanced liquid spray tightness
  • Very low inward leakage due to optimized design
  • Tyvek™ auto-lock zipper and storm flap for increased protection
  • Elastic face, wrists and ankles as well as glued-in waist elastic
Certifications
Chemical Protective Clothing, Cat.III,Type 5B,6B
Specifications
Specifications
Unisex
White
Elastic (Glued-in)
Elastic
Zipper
Non-sterile
None
Tyvek
Single-use
Elastic
Hooded
Serged
Chemicals
Storm Flap
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'