missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ Tyvek™ 500 Hooded Coverall
Description
- Pin locking slider locks in place when the pins on the puller are pushed into the zipper elements
- Products made after January, 2023 are manufactured under specifications that do not contain natural rubber latex.
- Protection against infective agents (EN 14126) including resistance to penetration by blood and body fluids using synthetic blood (ISO 16603 Class 3) and resistance to penetration by blood-borne pathogens using Phi-X174 bacteriophage (ISO 16604 Class 2)
- Chemical protective clothing, Category III, Type 5-B and 6-B. (1956)
- EN 1073-2 (protection against radioactive contamination)
- Antistatic treatment (EN 1149-5) - on both sides.
- Serged external seams featuring patent-pending technology for enhanced liquid spray tightness
- Very low inward leakage due to optimized design
- Tyvek™ auto-lock zipper and storm flap for increased protection
- Elastic face, wrists and ankles as well as glued-in waist elastic
Certifications
Chemical Protective Clothing, Cat.III,Type 5B,6B
Specifications
Specifications
| Product Type | Protective Coveralls |
| Size | 6X-Large |
| Gender | Unisex |
| Material | Tyvek |
| Color | White |
| Disposable | Single-use |
| Waist Style | Elastic (Glued-in) |
| Ankle Style | Elastic |
| Cuff Style | Elastic |
| Neck Style | Hooded |
| Show More |