missing translation for 'onlineSavingsMsg'
Learn More

Tyrphostin AG 835 98.0+%, TCI America™

Supplier:  TCI America T3540100MG

 View more versions of this product

Catalog No. T3540100MG


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Tyrphostin AG 835
146°C
MFCD00209855
tyrphostin b44, + enantiomer, --r-n-, a-methylbenzyl-3,4-dihydroxybenzylidenecyanoacetamide
CC(C1=CC=CC=C1)NC(=O)C(=CC2=CC(=C(C=C2)O)O)C#N
308.337
308.34
133550-37-5
C18H16N2O3
100 mg
UMGQVUWXNOJOSJ-CDCQXCROSA-N
(Z)-2-cyano-3-(3,4-dihydroxyphenyl)-N-[(1R)-1-phenylethyl]prop-2-enamide
23919058
≥98.0% (HPLC)

Chemical Identifiers

133550-37-5
308.337
UMGQVUWXNOJOSJ-CDCQXCROSA-N
23919058
CC(C1=CC=CC=C1)NC(=O)C(=CC2=CC(=C(C=C2)O)O)C#N
C18H16N2O3
MFCD00209855
tyrphostin b44, + enantiomer, --r-n-, a-methylbenzyl-3,4-dihydroxybenzylidenecyanoacetamide
(Z)-2-cyano-3-(3,4-dihydroxyphenyl)-N-[(1R)-1-phenylethyl]prop-2-enamide
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Freezer

SDS
Documents

Documents

Promotions

Promotions